Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAGCGGGCAGTGTGGCTCATAAG[A/C]CACGAACCGGGAACTCCACTTTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614368 MIM: 604469 | ||||||||||||||||||||
Literature Links: |
AP5M1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP5M1 - adaptor related protein complex 5 mu 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018229.3 | 387 | Missense Mutation | AGA,AGC | R,S 10 | NP_060699.3 | |
XM_006720196.2 | 387 | Intron | XP_006720259.1 | |||
XM_011536940.2 | 387 | Missense Mutation | AGA,AGC | R,S 10 | XP_011535242.1 |
EXOC5 - exocyst complex component 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006544.3 | 387 | Intron | NP_006535.1 | |||
XM_005267272.3 | 387 | Intron | XP_005267329.1 |