Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCAGGTTTTACAGTTTCTGGTA[C/T]AGGTTCTGCCTCTTTCTTTGTTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602984 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MED6 - mediator complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284209.1 | 693 | Missense Mutation | ATA,GTA | I,V 221 | NP_001271138.1 | |
NM_001284210.1 | 693 | Missense Mutation | TAT,TGT | Y,C 175 | NP_001271139.1 | |
NM_001284211.1 | 693 | Missense Mutation | ATA,GTA | I,V 214 | NP_001271140.1 | |
NM_005466.3 | 693 | Missense Mutation | ATA,GTA | I,V 214 | NP_005457.2 |