Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTATGATCGCATCAGGGATGTTGG[C/T]CCTGACAGGGCGGCATCCGAGTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603441 | ||||||||||||||||||||
Literature Links: |
ATP5S PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP5S - ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003803.2 | 766 | Silent Mutation | GGC,GGT | G,G 61 | NP_001003803.1 | |
NM_001003805.2 | 766 | Silent Mutation | GGC,GGT | G,G 61 | NP_001003805.1 | |
NM_015684.3 | 766 | Silent Mutation | GGC,GGT | G,G 61 | NP_056499.2 | |
XM_005267537.3 | 766 | Silent Mutation | GGC,GGT | G,G 46 | XP_005267594.1 | |
XM_011536649.2 | 766 | Silent Mutation | GGC,GGT | G,G 90 | XP_011534951.1 | |
XM_011536650.2 | 766 | Silent Mutation | GGC,GGT | G,G 90 | XP_011534952.1 | |
XM_011536651.2 | 766 | Silent Mutation | GGC,GGT | G,G 90 | XP_011534953.1 | |
XM_011536652.1 | 766 | Silent Mutation | GGC,GGT | G,G 51 | XP_011534954.1 | |
XM_011536653.2 | 766 | Silent Mutation | GGC,GGT | G,G 46 | XP_011534955.1 | |
XM_011536654.2 | 766 | Silent Mutation | GGC,GGT | G,G 90 | XP_011534956.1 | |
XM_011536655.2 | 766 | Silent Mutation | GGC,GGT | G,G 90 | XP_011534957.1 | |
XM_011536656.2 | 766 | Silent Mutation | GGC,GGT | G,G 61 | XP_011534958.1 | |
XM_011536657.2 | 766 | Silent Mutation | GGC,GGT | G,G 46 | XP_011534959.1 | |
XM_017021220.1 | 766 | Silent Mutation | GGC,GGT | G,G 46 | XP_016876709.1 |
CDKL1 - cyclin dependent kinase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |