Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTGGGCAGCTTGATATGCACACC[C/T]GCCCGCAGCCCGGTGCCCAGGTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123280 | ||||||||||||||||||||
Literature Links: |
CKB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CKB - creatine kinase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001823.4 | 1031 | Silent Mutation | GCA,GCG | A,A 293 | NP_001814.2 | |
XM_017020951.1 | 1031 | Silent Mutation | GCA,GCG | A,A 317 | XP_016876440.1 | |
XM_017020952.1 | 1031 | Intron | XP_016876441.1 |
TRMT61A - tRNA methyltransferase 61A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |