Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTACACTGGATGTGCCCAAAGCAG[A/G]GGCACAGCCCACAAAGCATGGCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 191050 | ||||||||||||||||||||
Literature Links: |
WARS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WARS - tryptophanyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR25 - WD repeat domain 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161476.1 | 282 | Missense Mutation | GAG,GGG | E,G 61 | NP_001154948.1 | |
NM_024515.4 | 282 | Missense Mutation | GAG,GGG | E,G 61 | NP_078791.3 | |
XM_005268056.4 | 282 | Missense Mutation | GAG,GGG | E,G 61 | XP_005268113.1 | |
XM_005268057.3 | 282 | Intron | XP_005268114.1 | |||
XM_005268058.4 | 282 | Intron | XP_005268115.1 | |||
XM_011537150.2 | 282 | Missense Mutation | GAG,GGG | E,G 61 | XP_011535452.1 | |
XM_011537151.2 | 282 | Intron | XP_011535453.2 | |||
XM_017021639.1 | 282 | Intron | XP_016877128.1 |