Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGCCCGAGATGGGGTGCAGGCC[C/T]AGGCCCAGGAAGGAGCTGGCCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610862 MIM: 616912 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEGS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DEGS2 - delta 4-desaturase, sphingolipid 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206918.2 | 748 | Silent Mutation | CTA,CTG | L,L 220 | NP_996801.2 | |
XM_006720043.3 | 748 | Silent Mutation | CTA,CTG | L,L 184 | XP_006720106.1 |
EVL - Enah/Vasp-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |