Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGGCCCAGGCCTACCTGGACCAC[C/G]TGTGGGATGAAGGCTGTGCCGTCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612498 MIM: 605567 | ||||||||||||||||||||
Literature Links: |
ADSSL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADSSL1 - adenylosuccinate synthase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIVA1 - SIVA1 apoptosis inducing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006427.3 | 238 | Missense Mutation | CTG,GTG | L,V 57 | NP_006418.2 | |
NM_021709.2 | 238 | Intron | NP_068355.1 | |||
XM_011536360.2 | 238 | Missense Mutation | CTG,GTG | L,V 57 | XP_011534662.1 |