Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCATAGTCTTACCTTAAAGGAA[A/G]TCTTTTTAATCTCTGGGTCCCCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GZMB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GZMB - granzyme B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004131.4 | 640 | Missense Mutation | ACT,ATT | T,I 197 | NP_004122.2 | |
XM_011536685.2 | 640 | Missense Mutation | ACT,ATT | T,I 185 | XP_011534987.1 |