Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCCGGATGGTCACCACCTCACCA[C/T]GCTTCACCACCAGACGGCTGTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMED8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMED8 - transmembrane p24 trafficking protein family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213601.1 | 459 | Missense Mutation | CAT,CGT | H,R 187 | NP_998766.1 | |
XM_017021223.1 | 459 | Missense Mutation | CAT,CGT | H,R 294 | XP_016876712.1 | |
XM_017021224.1 | 459 | Missense Mutation | CAT,CGT | H,R 124 | XP_016876713.1 | |
XM_017021225.1 | 459 | Missense Mutation | CAT,CGT | H,R 119 | XP_016876714.1 |