Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAGGCTTCAGCCGTCCTCTGCC[G/T]TTTAAGAGGTGGATCTTCATGATTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602659 MIM: 616518 MIM: 611023 | ||||||||||||||||||||
Literature Links: |
MNAT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MNAT1 - MNAT1, CDK activating kinase assembly factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC38A6 - solute carrier family 38 member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT5 - tRNA methyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020810.3 | 1668 | Silent Mutation | AGG,CGG | R,R 491 | NP_065861.3 | |
XM_005267916.3 | 1668 | Silent Mutation | AGG,CGG | R,R 519 | XP_005267973.2 | |
XM_011537017.2 | 1668 | Silent Mutation | AGG,CGG | R,R 518 | XP_011535319.1 |