Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCAAAGGCCATGAGGAAGTTGAG[C/T]TTTGCCGATGAGGTGACCACATCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614027 MIM: 608884 | ||||||||||||||||||||
Literature Links: |
INSM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INSM2 - INSM transcriptional repressor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032594.3 | 889 | Silent Mutation | AGC,AGT | S,S 226 | NP_115983.3 |
RALGAPA1 - Ral GTPase activating protein catalytic alpha subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |