Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTAAGTGTTATTTATCTCTCTAT[C/T]TTCGGCACCTAGCACATAGTAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609411 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COX16 - COX16, cytochrome c oxidase assembly homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNJ2BP - synaptojanin 2 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018373.2 | 5649 | UTR 3 | NP_060843.2 |
SYNJ2BP-COX16 - SYNJ2BP-COX16 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202547.2 | 5649 | Intron | NP_001189476.1 | |||
NM_001202548.2 | 5649 | Intron | NP_001189477.1 | |||
NM_001202549.2 | 5649 | Intron | NP_001189478.1 |