Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGTATGCCCCTCGCCAGGGGCC[A/T]TGAGGCCAAGCCCCAGGCACAGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605304 MIM: 607439 | ||||||||||||||||||||
Literature Links: |
NGB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NGB - neuroglobin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POMT2 - protein O-mannosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013382.5 | 2386 | Missense Mutation | AGG,TGG | R,W 695 | NP_037514.2 | |
XM_011536675.2 | 2386 | Missense Mutation | AGG,TGG | R,W 758 | XP_011534977.1 | |
XM_011536676.2 | 2386 | Missense Mutation | AGG,TGG | R,W 647 | XP_011534978.1 | |
XM_011536677.2 | 2386 | Missense Mutation | AGG,TGG | R,W 605 | XP_011534979.1 | |
XM_011536679.2 | 2386 | Missense Mutation | AGG,TGG | R,W 456 | XP_011534981.1 | |
XM_017021244.1 | 2386 | Missense Mutation | AGG,TGG | R,W 393 | XP_016876733.1 | |
XM_017021245.1 | 2386 | Missense Mutation | AGG,TGG | R,W 393 | XP_016876734.1 |