Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTCGGGCGTTCAGCCGCTCGCT[C/G]TCCGCCTTCCAGAAGGTACGGTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126063 MIM: 615094 | ||||||||||||||||||||
Literature Links: |
DLST PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLST - dihydrolipoamide S-succinyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244883.1 | 111 | Silent Mutation | CTC,CTG | L,L 16 | NP_001231812.1 | |
NM_001933.4 | 111 | Intron | NP_001924.2 |
PROX2 - prospero homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |