Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCAAGCCCCAGAGGACCTCGAGG[C/T]GCGTGTGGTTCCGAGAACCCCCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C14orf180 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C14orf180 - chromosome 14 open reading frame 180 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008404.2 | 538 | Missense Mutation | CGC,TGC | R,C 68 | NP_001008404.1 | |
NM_001286399.1 | 538 | Missense Mutation | CGC,TGC | R,C 68 | NP_001273328.1 | |
NM_001286400.1 | 538 | Missense Mutation | CGC,TGC | R,C 68 | NP_001273329.1 | |
XM_005267638.3 | 538 | Missense Mutation | CGC,TGC | R,C 68 | XP_005267695.1 | |
XM_011536764.2 | 538 | Missense Mutation | CGC,TGC | R,C 68 | XP_011535066.1 |
TMEM179 - transmembrane protein 179 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |