Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGGTGGTTTGATGTTCAAAAAT[A/G]TGATGGAAGCCGGGTGTGGTCTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTR10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTR10 - actin-related protein 10 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018477.2 | Intron | NP_060947.1 | ||||
XM_011536960.1 | Intron | XP_011535262.1 | ||||
XM_011536961.1 | Intron | XP_011535263.1 | ||||
XM_011536962.2 | Intron | XP_011535264.1 | ||||
XM_011536963.2 | Intron | XP_011535265.1 |