Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTGTCTGTGATTTGCTCAGCA[C/T]TGTCTGCAAAAATCGTCGGCACAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601981 | ||||||||||||||||||||
Literature Links: |
RNASE6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNASE6 - ribonuclease A family member k6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005615.4 | 541 | Missense Mutation | ACT,ATT | T,I 83 | NP_005606.1 | |
XM_017021566.1 | 541 | Missense Mutation | ACT,ATT | T,I 83 | XP_016877055.1 | |
XM_017021567.1 | 541 | Missense Mutation | ACT,ATT | T,I 83 | XP_016877056.1 |