Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAGCCAAACCACTTACCTTTCC[A/G]GCTTCATCTTCTCCCGCAGAGGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601121 | ||||||||||||||||||||
Literature Links: |
PGF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PGF - placental growth factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207012.1 | 938 | Missense Mutation | CCG,CTG | P,L 139 | NP_001193941.1 | |
NM_001293643.1 | 938 | Missense Mutation | CCG,CTG | P,L 138 | NP_001280572.1 | |
NM_002632.5 | 938 | Missense Mutation | CCG,CTG | P,L 139 | NP_002623.2 |