Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAGGCCGCCGCCCTCCATGCCCA[C/T]TTTCTTGTAGCTTTCCTCCAGGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600635 | ||||||||||||||||||||
Literature Links: |
NKX2-1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NKX2-1 - NK2 homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079668.2 | 420 | Missense Mutation | ATG,GTG | M,V 57 | NP_001073136.1 | |
NM_003317.3 | 420 | Missense Mutation | ATG,GTG | M,V 27 | NP_003308.1 |
NKX2-1-AS1 - NKX2-1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SFTA3 - surfactant associated 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |