Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGTCTCATGCCTGCTGTTAGAGG[C/T]GCTTCATTGTTCTCTTTATCCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116831 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GZMH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GZMH - granzyme H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270780.1 | 583 | Missense Mutation | CAC,CGC | H,R 181 | NP_001257709.1 | |
NM_001270781.1 | 583 | Missense Mutation | CAC,CGC | H,R 159 | NP_001257710.1 | |
NM_033423.4 | 583 | Missense Mutation | CAC,CGC | H,R 245 | NP_219491.1 | |
XM_011536683.2 | 583 | Missense Mutation | CAC,CGC | H,R 211 | XP_011534985.1 |