Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGGCATCTCTGTAGGTGGCTTGG[C/T]AGATGGCTCGACGGGCCAGATCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600306 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMB5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMB5 - proteasome subunit beta 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130725.1 | 1008 | Missense Mutation | TAC,TGC | Y,C 117 | NP_001124197.1 | |
NM_001144932.2 | 1008 | UTR 3 | NP_001138404.1 | |||
NM_002797.4 | 1008 | Missense Mutation | TAC,TGC | Y,C 220 | NP_002788.1 | |
XM_005267871.2 | 1008 | Intron | XP_005267928.1 |