Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGGTGAACTGTAGCTGCTCACC[A/G]AGCTGCTGAGAAGAGCATCAATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613401 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOXRED1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOXRED1 - NADP dependent oxidoreductase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VIPAS39 - VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193314.1 | 1459 | Missense Mutation | TCG,TTG | S,L 487 | NP_001180243.1 | |
NM_001193315.1 | 1459 | Missense Mutation | TCG,TTG | S,L 487 | NP_001180244.1 | |
NM_001193316.1 | 1459 | Missense Mutation | TCG,TTG | S,L 438 | NP_001180245.1 | |
NM_001193317.1 | 1459 | Missense Mutation | TCG,TTG | S,L 487 | NP_001180246.1 | |
NM_022067.3 | 1459 | Missense Mutation | TCG,TTG | S,L 487 | NP_071350.2 | |
XM_011537066.1 | 1459 | Missense Mutation | TCG,TTG | S,L 456 | XP_011535368.1 | |
XM_017021579.1 | 1459 | Missense Mutation | TCG,TTG | S,L 456 | XP_016877068.1 | |
XM_017021580.1 | 1459 | Intron | XP_016877069.1 | |||
XM_017021581.1 | 1459 | Intron | XP_016877070.1 |