Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAGGAGAATCAGTGGCTACGGG[A/C]TGAACTGGCCAACACGCAGCAGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600025 | ||||||||||||||||||||
Literature Links: |
KLC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLC1 - kinesin light chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130107.1 | 673 | Missense Mutation | GAT,GCT | D,A 122 | NP_001123579.1 | |
NM_005552.4 | 673 | Missense Mutation | GAT,GCT | D,A 122 | NP_005543.2 | |
NM_182923.3 | 673 | Missense Mutation | GAT,GCT | D,A 122 | NP_891553.2 |