Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612028 MIM: 600654 MIM: 602161 MIM: 612487 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMC9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EMC9 - ER membrane protein complex subunit 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FITM1 - fat storage inducing transmembrane protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR7703 - microRNA 7703 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSME1 - proteasome activator subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281528.1 | Intron | NP_001268457.1 | ||||
NM_001281529.1 | Intron | NP_001268458.1 | ||||
NM_006263.3 | Intron | NP_006254.1 | ||||
NM_176783.2 | Intron | NP_788955.1 |
PSME2 - proteasome activator subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF31 - ring finger protein 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |