Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGAGTTTTCACCGTTTACAAGCC[C/T]GTTTTCTGTGGCCTTTGCTTTGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615671 | ||||||||||||||||||||
Literature Links: |
SETD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SETD3 - SET domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032233.2 | 1669 | Missense Mutation | AGG,GGG | R,G 551 | NP_115609.2 | |
NM_199123.1 | 1669 | Intron | NP_954574.1 | |||
XM_005268127.3 | 1669 | Missense Mutation | AGG,GGG | R,G 452 | XP_005268184.1 | |
XM_011537231.2 | 1669 | Missense Mutation | AGG,GGG | R,G 551 | XP_011535533.2 | |
XM_011537232.2 | 1669 | Missense Mutation | AGG,GGG | R,G 551 | XP_011535534.1 | |
XM_011537233.2 | 1669 | Missense Mutation | AGG,GGG | R,G 493 | XP_011535535.1 | |
XM_011537234.1 | 1669 | Missense Mutation | AGG,GGG | R,G 452 | XP_011535536.1 | |
XM_011537235.1 | 1669 | Intron | XP_011535537.1 | |||
XM_017021699.1 | 1669 | Missense Mutation | AGG,GGG | R,G 551 | XP_016877188.1 | |
XM_017021700.1 | 1669 | Missense Mutation | AGG,GGG | R,G 551 | XP_016877189.1 | |
XM_017021701.1 | 1669 | Missense Mutation | AGG,GGG | R,G 493 | XP_016877190.1 | |
XM_017021702.1 | 1669 | Missense Mutation | AGG,GGG | R,G 394 | XP_016877191.1 |