Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCGGAGCAGCTCCGCGAGGACC[G/T]CACCATCCCGCTCATTGTGAGTACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615132 | ||||||||||||||||||||
Literature Links: |
GALNT16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GALNT16 - polypeptide N-acetylgalactosaminyltransferase 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168368.1 | 300 | Missense Mutation | CGC,CTC | R,L 54 | NP_001161840.1 | |
NM_020692.2 | 300 | Missense Mutation | CGC,CTC | R,L 54 | NP_065743.2 | |
XM_011537005.1 | 300 | Missense Mutation | CGC,CTC | R,L 54 | XP_011535307.1 | |
XM_011537006.2 | 300 | Missense Mutation | CGC,CTC | R,L 54 | XP_011535308.1 | |
XM_011537007.2 | 300 | Missense Mutation | CGC,CTC | R,L 54 | XP_011535309.1 | |
XM_017021498.1 | 300 | Missense Mutation | CGC,CTC | R,L 54 | XP_016876987.1 |
LOC105370550 - uncharacterized LOC105370550 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |