Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGGGAAGCATGTGAGATCAGCT[A/G]TCTTTGATCCCCTCCTTCTGTTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603534 MIM: 611612 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP1G2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JPH4 - junctophilin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146028.1 | 552 | UTR 3 | NP_001139500.1 | |||
NM_032452.2 | 552 | UTR 3 | NP_115828.2 |
LOC102724814 - uncharacterized LOC102724814 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THTPA - thiamine triphosphatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |