Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGAGCAAGAAAATTCAGTGCCAC[C/T]TTGGGTCTGTCTGCAATATACAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610511 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC23A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC23A - Sec23 homolog A, coat complex II component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006364.3 | 2932 | UTR 3 | NP_006355.2 | |||
XM_005267262.1 | 2932 | UTR 3 | XP_005267319.1 | |||
XM_011536355.2 | 2932 | Intron | XP_011534657.1 | |||
XM_017020928.1 | 2932 | Intron | XP_016876417.1 |