Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCTCAGAAAAGTGCCTCGAGCT[A/C]TTTGAGAGAAGGACTGTGTATGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD12B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABHD12B - abhydrolase domain containing 12B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206673.1 | Intron | NP_001193602.1 | ||||
NM_181533.3 | Intron | NP_853511.2 | ||||
NM_181814.1 | Intron | NP_861535.1 | ||||
XM_006720052.1 | Intron | XP_006720115.1 | ||||
XM_011536472.1 | Intron | XP_011534774.1 | ||||
XM_011536474.2 | Intron | XP_011534776.1 | ||||
XM_017021031.1 | Intron | XP_016876520.1 |