Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAGGGGGGCGAGGGCAGCTCAG[C/G]TGCGGGGCAGGCCGGGCCCTGGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603544 | ||||||||||||||||||||
Literature Links: |
CCDC85C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC85C - coiled-coil domain containing 85C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144995.1 | 1425 | Missense Mutation | CCT,GCT | P,A 332 | NP_001138467.1 | |
XM_011536705.2 | 1425 | Missense Mutation | CCT,GCT | P,A 345 | XP_011535007.1 | |
XM_011536706.2 | 1425 | Intron | XP_011535008.1 | |||
XM_011536707.2 | 1425 | Intron | XP_011535009.1 |
CCNK - cyclin K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |