Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGGACGTAGCCCTTCCTATAAAG[C/T]GATTCGTATTTACACGCTGGTATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614014 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNASE9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNASE9 - ribonuclease A family member 9 (inactive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001673.3 | 631 | Silent Mutation | NP_001001673.2 | |||
NM_001110356.1 | 631 | Silent Mutation | NP_001103826.1 | |||
NM_001110357.1 | 631 | Silent Mutation | NP_001103827.1 | |||
NM_001110358.1 | 631 | Silent Mutation | NP_001103828.1 | |||
NM_001110359.1 | 631 | Silent Mutation | NP_001103829.1 | |||
NM_001110360.1 | 631 | Silent Mutation | NP_001103830.1 | |||
NM_001110361.1 | 631 | Silent Mutation | NP_001103831.1 | |||
NM_001289110.1 | 631 | Silent Mutation | NP_001276039.1 |