Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACTGTTCCAACTGGCGATGCCCA[A/G]CTCGAGTGCAGGAGGATCAGCAGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C14orf28 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C14orf28 - chromosome 14 open reading frame 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017923.1 | 750 | Missense Mutation | ACT,GCT | T,A 162 | NP_001017923.1 | |
XM_011536408.1 | 750 | Intron | XP_011534710.1 |
LOC101927418 - uncharacterized LOC101927418 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |