Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGACCCCAGGCTCAGCCCAACAA[C/T]GCTTCACCCAGGTCTCCGATGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601931 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL2L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCL2L2 - BCL2 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199839.1 | 461 | Missense Mutation | CGC,TGC | R,C 78 | NP_001186768.1 | |
NM_004050.4 | 461 | Missense Mutation | CGC,TGC | R,C 78 | NP_004041.1 |
BCL2L2-PABPN1 - BCL2L2-PABPN1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199864.1 | 461 | Missense Mutation | CGC,TGC | R,C 78 | NP_001186793.1 |
PPP1R3E - protein phosphatase 1 regulatory subunit 3E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |