Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACACTTCTCTTCTCCCTGCGGA[A/T]CAACAGCATCCAGGGCTAGCAGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616003 MIM: 603885 | ||||||||||||||||||||
Literature Links: |
APOPT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOPT1 - apoptogenic 1, mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BAG5 - BCL2 associated athanogene 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015048.2 | 1672 | Missense Mutation | GAT,GTT | D,V 413 | NP_001015048.1 | |
NM_001015049.2 | 1672 | Missense Mutation | GAT,GTT | D,V 454 | NP_001015049.1 | |
NM_004873.3 | 1672 | Missense Mutation | GAT,GTT | D,V 413 | NP_004864.1 |