Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCACGAAAAGGCGCGGGGGCTGA[G/T]TAGCCTGGGTACTGGACATCAGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609906 MIM: 611461 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFS - embryonal Fyn-associated substrate | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277174.1 | 1492 | Missense Mutation | AAT,ACT | N,T 295 | NP_001264103.1 | |
NM_005864.3 | 1492 | Missense Mutation | AAT,ACT | N,T 464 | NP_005855.1 | |
NM_032459.2 | 1492 | Missense Mutation | AAT,ACT | N,T 371 | NP_115835.1 | |
XM_005267256.1 | 1492 | Missense Mutation | AAT,ACT | N,T 434 | XP_005267313.1 |
SLC22A17 - solute carrier family 22 member 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |