Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCAGCAAGAAGAGGCAAACTCA[C/T]TCTCTCACTGGATAAACAATCCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 153619 | ||||||||||||||||||||
Literature Links: |
DLGAP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLGAP5 - DLG associated protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146015.1 | 2387 | Missense Mutation | ATG,GTG | M,V 724 | NP_001139487.1 | |
NM_014750.4 | 2387 | Missense Mutation | ATG,GTG | M,V 724 | NP_055565.3 | |
XM_017021840.1 | 2387 | Missense Mutation | ATG,GTG | M,V 724 | XP_016877329.1 |
LGALS3 - lectin, galactoside binding soluble 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |