Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCACTCTTCAGAAATCCTTTTC[C/T]ATCTGATCCACATGGAGAGGTTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 167416 MIM: 607571 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAX9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAX9 - paired box 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A21 - solute carrier family 25 member 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171170.1 | 2818 | Intron | NP_001164641.1 | |||
NM_030631.3 | 2818 | UTR 3 | NP_085134.1 | |||
XM_011537287.2 | 2818 | Intron | XP_011535589.1 | |||
XM_011537288.2 | 2818 | Intron | XP_011535590.1 | |||
XM_011537289.2 | 2818 | Intron | XP_011535591.1 |