Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAGCAGCGGCCGCCATGGCCAA[A/G]CCCAGCGTGGAGCTCACCCGCGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614716 MIM: 615195 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARMIL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CARMIL3 - capping protein regulator and myosin 1 linker 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138360.3 | 166 | Silent Mutation | AAA,AAG | K,K 3 | NP_612369.3 | |
XM_011537300.2 | 166 | Intron | XP_011535602.1 | |||
XM_017021751.1 | 166 | Intron | XP_016877240.1 |
DHRS4L1 - dehydrogenase/reductase 4 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277864.1 | 166 | Intron | NP_001264793.1 | |||
XM_006720246.3 | 166 | Intron | XP_006720309.1 |