Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCAGTCGGGGATGTTCAAACGT[C/T]TGTCCATTCTGAGCTTTCACGCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602168 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VRK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VRK1 - vaccinia related kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003384.2 | Intron | NP_003375.1 | ||||
XM_006720247.3 | Intron | XP_006720310.1 | ||||
XM_011537132.1 | Intron | XP_011535434.1 | ||||
XM_017021622.1 | Intron | XP_016877111.1 | ||||
XM_017021624.1 | Intron | XP_016877113.1 | ||||
XM_017021625.1 | Intron | XP_016877114.1 | ||||
XM_017021626.1 | Intron | XP_016877115.1 |