Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGCTCCAGCCCGAGGCTCAGAA[C/T]GGGTACTGCGACACATATATCCGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604580 | ||||||||||||||||||||
Literature Links: |
FBLN5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBLN5 - fibulin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006329.3 | 1904 | Silent Mutation | CCA,CCG | P,P 447 | NP_006320.2 | |
XM_005267267.3 | 1904 | Silent Mutation | CCA,CCG | P,P 464 | XP_005267324.1 | |
XM_011536356.1 | 1904 | UTR 3 | XP_011534658.1 | |||
XM_011536357.1 | 1904 | UTR 3 | XP_011534659.1 | |||
XM_011536358.1 | 1904 | UTR 3 | XP_011534660.1 | |||
XM_017020929.1 | 1904 | Silent Mutation | CCA,CCG | P,P 391 | XP_016876418.1 |
TC2N - tandem C2 domains, nuclear | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |