Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGTAAATCTGGTTAGTGATGAC[C/T]AGGAGCAGGCGCTGAAGCTTTTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604573 | ||||||||||||||||||||
Literature Links: |
C14orf2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C14orf2 - chromosome 14 open reading frame 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127393.1 | 263 | Missense Mutation | AGT,GGT | S,G 73 | NP_001120865.1 | |
NM_004894.2 | 263 | Missense Mutation | AGT,GGT | S,G 56 | NP_004885.1 |