Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGCAAAGAGGTGAAAATTTTGTT[C/T]TGAAGTCTTAAGGAAACAGAGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610178 MIM: 607384 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0586 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0586 - KIAA0586 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244189.1 | Intron | NP_001231118.1 | ||||
NM_001244190.1 | Intron | NP_001231119.1 | ||||
NM_001244191.1 | Intron | NP_001231120.1 | ||||
NM_001244192.1 | Intron | NP_001231121.1 | ||||
NM_001244193.1 | Intron | NP_001231122.1 |
TIMM9 - translocase of inner mitochondrial membrane 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304485.1 | Intron | NP_001291414.1 | ||||
NM_001304486.1 | Intron | NP_001291415.1 | ||||
NM_001304487.1 | Intron | NP_001291416.1 | ||||
NM_001304488.1 | Intron | NP_001291417.1 | ||||
NM_001304489.1 | Intron | NP_001291418.1 | ||||
NM_001304490.1 | Intron | NP_001291419.1 | ||||
NM_001304491.1 | Intron | NP_001291420.1 | ||||
NM_012460.3 | Intron | NP_036592.1 |