Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAATCCTCCAGCAGCAGCTCCAG[C/T]TCCTCCAGCGGCAGGCGCACCCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602706 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRDE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NRDE2 - NRDE-2, necessary for RNA interference, domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017970.3 | 3704 | Silent Mutation | GAA,GAG | E,E 1157 | NP_060440.2 | |
XM_011536886.1 | 3704 | Silent Mutation | GAA,GAG | E,E 1167 | XP_011535188.1 | |
XM_017021394.1 | 3704 | Intron | XP_016876883.1 | |||
XM_017021395.1 | 3704 | Silent Mutation | GAA,GAG | E,E 926 | XP_016876884.1 | |
XM_017021396.1 | 3704 | Silent Mutation | GAA,GAG | E,E 662 | XP_016876885.1 |
PSMC1 - proteasome 26S subunit, ATPase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |