Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCGGGAGGACGCACTGAGGTACAC[A/G]CTGGACTTCCTGGTACCAGCCAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610018 MIM: 605272 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF40 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF40 - Rho guanine nucleotide exchange factor 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278529.1 | 319 | UTR 5 | NP_001265458.1 | |||
NM_001278530.1 | 319 | UTR 5 | NP_001265459.1 | |||
NM_018071.4 | 319 | Silent Mutation | ACA,ACG | T,T 48 | NP_060541.3 | |
XM_005267844.2 | 319 | Silent Mutation | ACA,ACG | T,T 48 | XP_005267901.1 | |
XM_011536937.2 | 319 | Silent Mutation | ACA,ACG | T,T 48 | XP_011535239.1 | |
XM_011536938.2 | 319 | Silent Mutation | ACA,ACG | T,T 48 | XP_011535240.1 | |
XM_017021434.1 | 319 | UTR 5 | XP_016876923.1 | |||
XM_017021435.1 | 319 | UTR 5 | XP_016876924.1 | |||
XM_017021436.1 | 319 | Intron | XP_016876925.1 | |||
XM_017021437.1 | 319 | Intron | XP_016876926.1 |
NDRG2 - NDRG family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |