Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTCGCCTCCCTCGGCTCGGCCCA[A/G]AGCAGCAGCTTCAGCCCCGAAGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600754 MIM: 611856 | ||||||||||||||||||||
Literature Links: |
MMP14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MMP14 - matrix metallopeptidase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004995.3 | 372 | Silent Mutation | CAA,CAG | Q,Q 29 | NP_004986.1 |
MRPL52 - mitochondrial ribosomal protein L52 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |