Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCCCCATCATGCCTGTCAGTAC[C/T]GTGTCCAGGTCGTAGTAGGGGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612270 | ||||||||||||||||||||
Literature Links: |
CDCA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDCA4 - cell division cycle associated 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017955.3 | 675 | Silent Mutation | ACA,ACG | T,T 193 | NP_060425.2 | |
NM_145701.2 | 675 | Silent Mutation | ACA,ACG | T,T 193 | NP_663747.1 |