Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCACAATTCTTCACGCTCCTTTT[C/G]TTTCTTTTTCTCACTGAGGAAGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601647 | ||||||||||||||||||||
Literature Links: |
PPP2R5E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP2R5E - protein phosphatase 2 regulatory subunit B'epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282179.1 | 1908 | Missense Mutation | CAA,GAA | Q,E 440 | NP_001269108.1 | |
NM_001282180.1 | 1908 | Missense Mutation | CAA,GAA | Q,E 435 | NP_001269109.1 | |
NM_001282181.1 | 1908 | Missense Mutation | CAA,GAA | Q,E 364 | NP_001269110.1 | |
NM_001282182.1 | 1908 | Missense Mutation | CAA,GAA | Q,E 364 | NP_001269111.1 | |
NM_006246.3 | 1908 | Missense Mutation | CAA,GAA | Q,E 440 | NP_006237.1 |