Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCAGGCTGGGTGACTGATGGGGA[C/T]GGGCAGCCTGAGGGTGGCCTGCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603728 | ||||||||||||||||||||
Literature Links: |
NUMB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUMB - NUMB, endocytic adaptor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005743.1 | 1933 | Missense Mutation | CAT,CGT | H,R 549 | NP_001005743.1 | |
NM_001005744.1 | 1933 | Missense Mutation | CAT,CGT | H,R 501 | NP_001005744.1 | |
NM_001005745.1 | 1933 | Missense Mutation | CAT,CGT | H,R 490 | NP_001005745.1 | |
NM_001320114.1 | 1933 | Missense Mutation | CAT,CGT | H,R 501 | NP_001307043.1 | |
NM_003744.5 | 1933 | Missense Mutation | CAT,CGT | H,R 538 | NP_003735.3 |
PAPLN - papilin, proteoglycan like sulfated glycoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |