Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCATGGCCGTCATGGACGCGTA[C/T]CTGGTGGAGCAGAACCAGGGCCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC105370697 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105370697 - uncharacterized LOC105370697 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM121 - transmembrane protein 121 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025268.2 | 842 | Silent Mutation | TAC,TAT | Y,Y 28 | NP_079544.1 | |
XM_005268101.3 | 842 | Silent Mutation | TAC,TAT | Y,Y 28 | XP_005268158.1 | |
XM_006720261.3 | 842 | Silent Mutation | TAC,TAT | Y,Y 28 | XP_006720324.1 |