Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCGGTCCCTTTCCCTATCCCGA[C/T]CTCGGTCCCCCCTGTCCCGCTCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604562 | ||||||||||||||||||||
Literature Links: |
ACIN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACIN1 - apoptotic chromatin condensation inducer 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164814.1 | 4037 | Missense Mutation | GAT,GGT | D,G 1290 | NP_001158286.1 | |
NM_001164815.1 | 4037 | Missense Mutation | GAT,GGT | D,G 1263 | NP_001158287.1 | |
NM_001164816.1 | 4037 | Missense Mutation | GAT,GGT | D,G 576 | NP_001158288.1 | |
NM_001164817.1 | 4037 | Missense Mutation | GAT,GGT | D,G 545 | NP_001158289.1 | |
NM_014977.3 | 4037 | Missense Mutation | GAT,GGT | D,G 1303 | NP_055792.1 | |
XM_005267415.3 | 4037 | Missense Mutation | GAT,GGT | D,G 1302 | XP_005267472.1 | |
XM_005267416.3 | 4037 | Missense Mutation | GAT,GGT | D,G 1291 | XP_005267473.1 | |
XM_005267418.1 | 4037 | Missense Mutation | GAT,GGT | D,G 545 | XP_005267475.1 | |
XM_006720081.3 | 4037 | Intron | XP_006720144.1 | |||
XM_011536570.2 | 4037 | Intron | XP_011534872.1 |
CDH24 - cadherin 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105370705 - collagen alpha-2(I) chain-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011537450.1 | 4037 | Intron | XP_011535752.1 |